Skip to Content
MilliporeSigma
All Photos(1)

Documents

412155

Sigma-Aldrich

Neopentyl glycol methyl ether propoxylate (2PO/OH) acrylate

average Mn ~288, contains 250 ppm monomethyl ether hydroquinone as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(C3H6O)nCH2C(CH3)2CH2OCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~288

contains

250 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.439 (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

COCC(C)(C)COCC(C)OCC(C)OC(=O)C=C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service