Skip to Content
MilliporeSigma
All Photos(1)

Documents

411973

Sigma-Aldrich

1,2,3-Trichloro-5-nitrobenzene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2NO2
CAS Number:
Molecular Weight:
226.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

68-71 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(Cl)c(Cl)c(Cl)c1

InChI

1S/C6H2Cl3NO2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H

InChI key

HHLCSFGOTLUREE-UHFFFAOYSA-N

General description

1,2,3-Trichloro-5-nitrobenzene is a chloronitrobenzene (CNB) compound. In vitro nephrotoxic potential of 1,2,3-trichloro-5-nitrobenzene in animal models and human kidney cells has been investigated. Toxicological study of 1,2,3-trichloro-5-nitrobenzene has been reported.2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(U) cracked the code: a Marshall Nirenberg memoir.
Robert Haselkorn
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(4), 968-969 (2010-04-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service