Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

411078

Sigma-Aldrich

1,1,2,2,3-Pentafluoropropane

99%

Synonym(s):

HFC-245ca

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FCH2CF2CHF2
CAS Number:
Molecular Weight:
134.05
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

40.81 psi ( 55 °C)

Assay

99%

refractive index

n20/D 1.567 (lit.)

bp

25-26 °C (lit.)

mp

−82 °C (lit.)

density

1.398 g/mL at 25 °C (lit.)

SMILES string

FCC(F)(F)C(F)F

InChI

1S/C3H3F5/c4-1-3(7,8)2(5)6/h2H,1H2

InChI key

AWTOFSDLNREIFS-UHFFFAOYSA-N

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service