Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41030

Sigma-Aldrich

4,4-Dimethyloxazolidine-2-thione

≥98.0% (CHN)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NOS
CAS Number:
Molecular Weight:
131.20
Beilstein:
109195
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (CHN)

mp

122-124 °C

SMILES string

CC1(C)COC(=S)N1

Other Notes

Starting material for the synthesis of achiral thiiranes

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.I. Meyers et al.
The Journal of Organic Chemistry, 41, 1735-1735 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service