Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

410004

Sigma-Aldrich

tert-Pentyl vinyl ether

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2OCH=CH2
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.411 (lit.)

bp

106 °C (lit.)

density

0.783 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)OC=C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service