Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

409618

Sigma-Aldrich

Bis[bis(trimethylsilyl)amino]tin(II)

Synonym(s):

Bis(bis(trimethylsilyl)amino)stannylene, Tin bis[bis(trimethylsilyl)amide], Tin(II) bis(hexamethyldisilazide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)3Si]2N]2Sn
CAS Number:
Molecular Weight:
439.48
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: tin

refractive index

n20/D 1.514 (lit.)

bp

112 °C/0.05 mmHg (lit.)

mp

37-38 °C (lit.)

density

1.136 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)N([SnH2]N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/2C6H18NSi2.Sn/c2*1-8(2,3)7-9(4,5)6;/h2*1-6H3;/q2*-1;+2

InChI key

WLNIUEYAQZRJFS-UHFFFAOYSA-N

Application

Reagent for the conversion of aldehydes and ketones to N,N-dialkyleneamines.[1][2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 57, 5063-5063 (1992)
Synlett, 131-131 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service