Skip to Content
MilliporeSigma
All Photos(1)

Documents

409529

Sigma-Aldrich

Poly(ethylene glycol) methacrylate

average Mn 500, methacrylate, 900 ppm MEHQ as inhibitor

Synonym(s):

Ethoxylated 2-hydroxyethyl methacrylate, Poly(oxy-1,2-ethanediyl), α-(2-methyl-1-oxo-2-propenyl)-ω-hydroxy-, Polyethylene glycol, Polyethylene glycol monomethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) methacrylate, average Mn 500, contains 900 ppm monomethyl ether hydroquinone as inhibitor

form

liquid

Quality Level

mol wt

average Mn 500

contains

900 ppm monomethyl ether hydroquinone as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.467

density

1.101 g/mL at 25 °C

Ω-end

hydroxyl

α-end

methacrylate

polymer architecture

shape: linear
functionality: heterobifunctional

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May darken upon storage.

Application

Comonomer in emulsion polymerization, thickener and dispersant and suspending aid.

Features and Benefits

Copolymerizable, non-ionic stabilizer. Hydrophilic, modifying comonomer and cross-linking agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary M Geisterfer et al.
Current biology : CB, 30(15), 3016-3023 (2020-06-13)
The microtubule cytoskeleton plays critically important roles in numerous cellular functions in eukaryotes, and it does so across a functionally diverse and morphologically disparate range of cell types [1]. In these roles, microtubule assemblies must adopt distinct morphologies and physical
Yanlu Chen et al.
Cell reports, 33(5), 108349-108349 (2020-11-05)
We present a tiling light sheet microscope compatible with all tissue clearing methods for rapid multicolor 3D imaging of cleared tissues with micron-scale (4 × 4 × 10 μm3) to submicron-scale (0.3 × 0.3 × 1 μm3) spatial resolution. The resolving ability is improved to sub-100 nm

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service