Skip to Content
MilliporeSigma
All Photos(1)

Documents

409146

Sigma-Aldrich

Poly(dimethylamine-co-epichlorohydrin-co-ethylenediamine) solution

average Mw ~250,000, 50 wt. % in H2O

Synonym(s):

Dimethylamine-ethylenediamine-epichlorohydrin copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mw ~250,000

concentration

50 wt. % in H2O

refractive index

n20/D 1.438

density

1.14 g/mL at 25 °C

SMILES string

CNC.NCCN.ClCC1CO1

InChI

1S/C3H5ClO.C2H8N2.C2H7N/c4-1-3-2-5-3;3-1-2-4;1-3-2/h3H,1-2H2;1-4H2;3H,1-2H3

InChI key

RPKDEWATZPCEIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service