Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

408956

Sigma-Aldrich

Isodecyl acrylate

contains MEHQ as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2C10H21
CAS Number:
Molecular Weight:
212.33
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

contains

MEHQ as inhibitor

refractive index

n20/D 1.442 (lit.)

bp

121 °C/10 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCCCOC(=O)C=C

InChI

1S/C13H24O2/c1-4-13(14)15-11-9-7-5-6-8-10-12(2)3/h4,12H,1,5-11H2,2-3H3

InChI key

LVGFPWDANALGOY-UHFFFAOYSA-N

Application

IDA monomers were doped with silica and gold nanoparticles for the preparation of amorphous chalcogenide-AuNP nanocomposite. [1],[2]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement of laser recording in gold/amorphous chalcogenide and gold/acrylate nanocomposite layers.
Burunkova J, et al.
Journal of Non-Crystalline Solids, 402, 200-203 (2014)
Ilana Talior-Volodarsky et al.
Biomaterials, 144, 199-210 (2017-08-26)
The IGF-1 signaling pathway and IGF-1-dependent macrophage/endothelial cell crosstalk was found to be critical features of the vascular regenerative effect displayed by implanted methacrylic acid -co-isodecyl acrylate (MAA-co-IDA; 40% MAA) coated disks in CD1 mice. Inhibition of IGF-1 signaling using
Influence of gold nanoparticles on the photo-polymerization processes and structure in acrylate nanocomposites.
Burunkova J, et al.
European Polymer Journal, 64, 189-195 (2015)
Application of a cold plasma process for polymerization and copolymerization of fluorinated and hydrogenated (meth) acrylates.
Hochart F, et al.
Polymer, 41(9), 3159-3166 (2000)
Qiji Ze et al.
Advanced materials (Deerfield Beach, Fla.), 32(4), e1906657-e1906657 (2019-12-10)
Shape-programmable soft materials that exhibit integrated multifunctional shape manipulations, including reprogrammable, untethered, fast, and reversible shape transformation and locking, are highly desirable for a plethora of applications, including soft robotics, morphing structures, and biomedical devices. Despite recent progress, it remains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service