Skip to Content
MilliporeSigma
All Photos(1)

Documents

40875

Sigma-Aldrich

(−)-2,3-O-Benzylidene-1,1,4,4-tetraphenyl-L-threitol polymer-bound

extent of labeling: ~0.4 mmol/g loading

Synonym(s):

(4R,5R)-2,α,α,α′,α′-Pentaphenyl-dioxolane-4,5-dimethanol polymer-bound, (R,R)-Taddol (phenyl) polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

extent of labeling

~0.4 mmol/g loading

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

The supported chiral titanate complex is employed as asymmetric catalyst in a batch reactor for enantioselective additions of alkyl groups to aldehydes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.J. Comina et al.
Organic Process Research & Development, 2, 18-18 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service