Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

408603

Sigma-Aldrich

Z-Glu-OMe

98%

Synonym(s):

N-Carbobenzyloxy-L-glutamic acid 1-methyl ester, Z-L-Glutamic acid 1-methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2CH(NHCOOCH2C6H5)COOCH3
CAS Number:
Molecular Weight:
295.29
Beilstein:
2567599
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −24°, c = 2 in ethanol

mp

68-70 °C (lit.)

SMILES string

COC(=O)[C@H](CCC(O)=O)NC(=O)OCc1ccccc1

InChI

1S/C14H17NO6/c1-20-13(18)11(7-8-12(16)17)15-14(19)21-9-10-5-3-2-4-6-10/h2-6,11H,7-9H2,1H3,(H,15,19)(H,16,17)/t11-/m0/s1

InChI key

BGMCTGARFXPQML-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service