Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

408425

Sigma-Aldrich

Boc-Met-OH

99%

Synonym(s):

N-(tert-Butoxycarbonyl)-L-methionine, Boc-L-methionine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$84.30
25 G
$156.00

$84.30


Available to ship onApril 08, 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$84.30
25 G
$156.00

About This Item

Linear Formula:
CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
249.33
Beilstein:
1727869
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$84.30


Available to ship onApril 08, 2025Details


Request a Bulk Order

Assay

99%

optical activity

[α]20/D −22°, c = 1 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

47-50 °C (lit.)

application(s)

peptide synthesis

SMILES string

CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C10H19NO4S/c1-10(2,3)15-9(14)11-7(8(12)13)5-6-16-4/h7H,5-6H2,1-4H3,(H,11,14)(H,12,13)/t7-/m0/s1

InChI key

IMUSLIHRIYOHEV-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shadab Alam et al.
International journal of nanomedicine, 7, 4207-4222 (2012-08-24)
Curcumin, the principal curcuminoid of the popular Indian spice turmeric, has a wide spectrum of pharmaceutical properties such as antitumor, antioxidant, antiamyloid, and anti-inflammatory activity. However, poor aqueous solubility and low bioavailability of curcumin is a major challenge in its
Edit Brodszkij et al.
Small (Weinheim an der Bergstrasse, Germany), 16(27), e1906493-e1906493 (2020-05-30)
Polymer-lipid hybrid vesicles are an emerging type of nano-assemblies that show potential as artificial organelles among others. Phospholipids and poly(cholesteryl methacrylate)-block-poly(methionine methacryloyloxyethyl ester (METMA)-random-2-carboxyethyl acrylate (CEA)) labeled with a Förster resonance energy transfer (FRET) reporter pair are used for the
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service