Skip to Content
MilliporeSigma
All Photos(1)

Documents

408344

Sigma-Aldrich

Tridecyl methacrylate

Mixture of C12-C14 alkyl esters

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2R, R = C12-C14
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

contains

600 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.45 (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCOC(=O)C(C)=C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service