Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407925

Sigma-Aldrich

4-Chloro-3-nitrobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)COCl
CAS Number:
Molecular Weight:
220.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

47-54 °C (lit.)

functional group

acyl chloride
chloro
nitro

SMILES string

[O-][N+](=O)c1cc(ccc1Cl)C(Cl)=O

InChI

1S/C7H3Cl2NO3/c8-5-2-1-4(7(9)11)3-6(5)10(12)13/h1-3H

InChI key

IWLGXPWQZDOMSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-3-nitrobenzoyl chloride is an acid halide.

Application

4-Chloro-3-nitrobenzoyl chloride may be employed as acylation reagent for the Friedel-Crafts acylation of activated benzenes such as anisole, veratrole and 1,4-dimethoxybenzene.[1] It may be employed for the synthesis of 4-chloro-4-methoxy-3-nitrobenzophenone.[1] It may be employed as acylation reagent for the acylation reaction of the deactivated amines (2- aminopyrimidine, aminopyrazine).[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzvetomira Tzanova et al.
European journal of medicinal chemistry, 44(6), 2724-2730 (2008-10-28)
Considering that oxidative stress is strongly implicated in the toxicity of chemotherapy, much effort is focused on the research of diverse antioxidants as protective agents. An efficient synthesis of three novel benzophenones containing 1,3-thiazol moiety (6a-c) is described. Their antioxidant
Insights into the N,N-diacylation reaction of 2-aminopyrimidines and deactivated anilines: an alternative N-monoacylation reaction.
Theodorou V, et al.
ARKIVOC (Gainesville, FL, United States), 4, 11-23 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service