Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407135

Sigma-Aldrich

(1R,2S)-(+)-2-(Dibutylamino)-1-phenyl-1-propanol

97%

Synonym(s):

(+)-DBNE, N,N-Dibutyl-L-(+)-norephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2NCH(CH3)CH(C6H5)OH
CAS Number:
Molecular Weight:
263.42
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +21°, c = 2 in chloroform

refractive index

n20/D 1.501 (lit.)

bp

121 °C/0.1 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CCCCN(CCCC)[C@@H](C)[C@H](O)c1ccccc1

InChI

1S/C17H29NO/c1-4-6-13-18(14-7-5-2)15(3)17(19)16-11-9-8-10-12-16/h8-12,15,17,19H,4-7,13-14H2,1-3H3/t15-,17-/m0/s1

InChI key

BRRGNOFUBFINSX-RDJZCZTQSA-N

General description

(1R,2S)-(+)-2-(Dibutylamino)-1-phenyl-1-propanol, a norephedrine derivative, is a chiral auxiliary for asymmetric synthesis.

Application

(+)-DBNE reacts with nickel bromide (NiBr2) to form a chiral catalyst, which can catalyze the enantioselective conjugate addition of dialkylzincs to α,β-unsaturated ketones leading to the formation of optically active β-substituted ketones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral N, N-dialkylnorephedrines as catalysts of the highly enantioselective addition of dialkylzincs to aliphatic and aromatic aldehydes. The asymmetric synthesis of secondary aliphatic and aromatic alcohols of high optical purity.
Soai K, et al.
The Journal of Organic Chemistry, 56(13), 4264-4268 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service