Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407054

Sigma-Aldrich

N,N-Dimethylacetoacetamide solution

80% in H2O

Synonym(s):

N,N-Dimethyl-3-oxobutyramide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CON(CH3)2
CAS Number:
Molecular Weight:
129.16
Beilstein:
1755038
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

concentration

80% in H2O

refractive index

n20/D 1.447

density

1.067 g/mL at 25 °C

SMILES string

CN(C)C(=O)CC(C)=O

InChI

1S/C6H11NO2/c1-5(8)4-6(9)7(2)3/h4H2,1-3H3

InChI key

YPEWWOUWRRQBAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service