40652
(S)-7,7′-Bis(diphenylphosphino)-3,3′,4,4′-tetrahydro-4,4′-dimethyl-8,8′-bi(2H-1,4-benzoxazine)
≥95.0%
Synonym(s):
Solphos SL-A001-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C42H38N2O2P2
CAS Number:
Molecular Weight:
664.71
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Assay
≥95.0%
optical activity
[α]20/D -137±10°, c = 0.5 in chloroform
optical purity
ee: ≥99%
SMILES string
CN1CCOc2c1ccc(P(c3ccccc3)c4ccccc4)c2-c5c6OCCN(C)c6ccc5P(c7ccccc7)c8ccccc8
General description
sold in collaboration with Solvias AG
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service