Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40617

Sigma-Aldrich

(2R,6R)-2,6-Heptanediol

≥99.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O2
CAS Number:
Molecular Weight:
132.20
Beilstein:
6954914
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

≥99.0% (sum of enantiomers, GC)

form

solid

optical purity

enantiomeric ratio: ≥99:1% (GC)

SMILES string

C[C@@H](O)CCC[C@@H](C)O

InChI

1S/C7H16O2/c1-6(8)4-3-5-7(2)9/h6-9H,3-5H2,1-2H3/t6-,7-/m1/s1

InChI key

OZIMXLFSRSPFAS-RNFRBKRXSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service