Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

406066

Sigma-Aldrich

Bis(3,4-epoxycyclohexylmethyl) adipate

Synonym(s):

Di(3,4-epoxycyclohexylmethyl) adipate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O6
CAS Number:
Molecular Weight:
366.45
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

refractive index

n20/D 1.493 (lit.)

density

1.149 g/mL at 25 °C (lit.)

SMILES string

O=C(CCCCC(=O)OCC1CCC2OC2C1)OCC3CCC4OC4C3

InChI

1S/C20H30O6/c21-19(23-11-13-5-7-15-17(9-13)25-15)3-1-2-4-20(22)24-12-14-6-8-16-18(10-14)26-16/h13-18H,1-12H2

InChI key

DJUWPHRCMMMSCV-UHFFFAOYSA-N

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service