Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

405388

Sigma-Aldrich

2-Chloro-4,5-difluorobenzoic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(F)2CO2H
CAS Number:
Molecular Weight:
192.55
Beilstein:
4247522
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

103-106 °C (lit.)

functional group

carboxylic acid
chloro
fluoro

SMILES string

OC(=O)c1cc(F)c(F)cc1Cl

InChI

1S/C7H3ClF2O2/c8-4-2-6(10)5(9)1-3(4)7(11)12/h1-2H,(H,11,12)

InChI key

CGFMLBSNHNWJAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-4,5-difluorobenzoic acid is an important intermediate in medicinal compounds and pesticides. Lanthanide complexes with the 2-chloro-4,5-difluorobenzoate (2-cl-4,5-dfba) and 1,10-phenanthroline (phen), have been prepared and their crystal structures have been studied. Efficient synthesis of 2-chloro-4,5-difluorobenzoic acid v-difluororbenzene has been reported.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structures and thermodynamic properties of lanthanide complexes with 2-chloro-4, 5-difluorobenzoate and 1, 10-phenanthroline.
Tang K, et al.
The Journal of Chemical Thermodynamics, 56, 38-48 (2013)
Syntheses of 2-Chloro-4, 5-difluorobenzoic Acid.
Li Z and Huang N.
Organic preparations and procedures international, 28(2), 245-246 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service