Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

405310

Sigma-Aldrich

Caprolactone 2-(methacryloyloxy)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)5CO2CH2CH2O2CC(CH3)=CH2
CAS Number:
Molecular Weight:
244.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

contains

200 ppm 2-methylhydroquinone as inhibitor

refractive index

n20/D 1.463 (lit.)

mp

−48 °C (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCOC(=O)CCCCCO

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ursula Pérez-Ramírez et al.
Computer methods in biomechanics and biomedical engineering, 18(4), 446-455 (2013-08-21)
Anisotropy is one of the most meaningful determinants of biomechanical behaviour. This study employs micro-computed tomography (μCT) and image techniques for analysing the anisotropy of regenerative medicine polymer scaffolds. For this purpose, three three-dimensional anisotropy evaluation image methods were used:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service