Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

405299

Sigma-Aldrich

m-Tolylmagnesium chloride solution

1.0 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4MgCl
CAS Number:
Molecular Weight:
150.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

density

0.956 g/mL at 25 °C

SMILES string

Cc1cccc([Mg]Cl)c1

InChI

1S/C7H7.ClH.Mg/c1-7-5-3-2-4-6-7;;/h2-3,5-6H,1H3;1H;/q;;+1/p-1

InChI key

BVYCQPCLCZTSPD-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-5.8 °F - closed cup

Flash Point(C)

-21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service