Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

405272

Sigma-Aldrich

Thallium(I) hexafluoroacetylacetonate

Synonym(s):

(Hexafluoroacetylacetonato)thallium(I)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlC5HF6O2
CAS Number:
Molecular Weight:
411.43
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: thallium
reagent type: catalyst

SMILES string

FC(F)(F)C(=O)\C=C(/O[Tl])C(F)(F)F

InChI

1S/C5H2F6O2.Tl/c6-4(7,8)2(12)1-3(13)5(9,10)11;/h1,12H;/q;+1/p-1/b2-1-;

InChI key

YHQMYQKZMHUWEZ-ODZAUARKSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service