Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40451

Sigma-Aldrich

Bis(tert-butylcyclohexyl) peroxydicarbonate

technical, ≥90% (RT)

Synonym(s):

Perkadox 16

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38O6
CAS Number:
Molecular Weight:
398.53
Beilstein:
2176683
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (RT)

form

solid

impurities

≥3% Oxygen (active)

storage temp.

2-8°C

SMILES string

CC(C)(C)C1CCC(CC1)OC(=O)OOC(=O)OC2CCC(CC2)C(C)(C)C

InChI

1S/C22H38O6/c1-21(2,3)15-7-11-17(12-8-15)25-19(23)27-28-20(24)26-18-13-9-16(10-14-18)22(4,5)6/h15-18H,7-14H2,1-6H3

InChI key

NOBYOEQUFMGXBP-UHFFFAOYSA-N

Application

Initiator for the polymerization of acrylic esters

Storage and Stability

Warning: these products are subject to the Explosives Act and must be transported refrigerated - additional costs for transport will apply.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Org. Perox. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Buijs et al.
Stain technology, 58(3), 135-141 (1983-05-01)
An improved routine method for embedding tissue, especially hard tissue, in polymethyl methacrylate (pMMA) is described. The improvements were: the final dehydration step before MMA infiltration was performed with methanol in a Soxhlet apparatus; the stabilizer hydroquinone was not extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service