Skip to Content
MilliporeSigma
All Photos(1)

Documents

404349

Sigma-Aldrich

[Bis(trifluoroacetoxy)iodo]pentafluorobenzene

97%

Synonym(s):

FPIFA, Iodopentafluorobenzene bis(trifluoroacetate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO2)2IC6F5
CAS Number:
Molecular Weight:
519.99
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reagent type: oxidant

mp

117-119 °C (lit.)

SMILES string

Fc1c(F)c(F)c([I](OC(=O)C(F)(F)F)OC(=O)C(F)(F)F)c(F)c1F

InChI

1S/C10F11IO4/c11-1-2(12)4(14)6(5(15)3(1)13)22(25-7(23)9(16,17)18)26-8(24)10(19,20)21

InChI key

OQWAXRPJEPTTSZ-UHFFFAOYSA-N

Application

Reactant or reagent for:
  • Oxidation of organic iodides using oxone and trifluoroacetic acid
  • Base hydrolysis
  • Oxidative cross-coupling of arenes
  • Oxidation of phenolic compounds with organohypervalent iodine
  • Aziridination of alkenes
  • Oxidation of organic compounds
  • Oxidative fragmentation of α-amino acids or β-amino alcohols
  • Oxidative biaryl coupling of thiophenes

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service