Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40280

Sigma-Aldrich

Nitric acid

semiconductor grade MOS PURANAL (Honeywell 17920), ≥69%

Synonym(s):

Nital

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

semiconductor grade MOS PURANAL (Honeywell 17920)

vapor pressure

8 mmHg ( 20 °C)

Assay

≥69%

CofA

specification on request

color

colorless

bp

120.5 °C (lit.)

density

1.400 g/cm3 at 25 °C

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole McGowan et al.
The Science of the total environment, 494-495, 177-186 (2014-07-22)
Microchemical analysis of otolith (calcified 'ear stones' used for balance and orientation) of fishes is an important tool for studying their environmental history and management. However, the spatial resolution achieved is often too coarse to examine short-term events occurring in
Alyson M Baergen et al.
Environmental science & technology, 47(2), 815-820 (2012-12-15)
The fate of NO(x) (=NO + NO(2)) is important to understand because NO(x) is a significant player in air quality determination through its role in O(3) formation. Here we show that renoxification of the urban atmosphere may occur through the
Lauren E Kremer et al.
Journal of inorganic biochemistry, 147, 227-234 (2015-05-11)
Uncontrolled reactions in biological media are a main obstacle for clinical translation of V-based anti-diabetic or anti-cancer pro-drugs. We investigated the use of controlled-release pharmaceutical formulations to ameliorate this issue with a series of V(V) and (IV) complexes of anionic
Paul A Adlard et al.
Neurobiology of disease, 81, 196-202 (2015-01-01)
Zinc transporter-3 (ZnT3) protein is responsible for loading zinc into presynaptic vesicles and consequently controls the availability of zinc at the glutamatergic synapse. ZnT3 has been shown to decline with age and in Alzheimer's disease (AD) and is crucially involved
Stijn Smulders et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(1), 132-140 (2014-06-14)
The unique physical and chemical properties of nanomaterials have led to their increased use in many industrial applications, including as a paint additive. For example, titanium dioxide (TiO2) engineered nanoparticles (ENPs) have well-established anti-UV, self-cleaning, and air purification effects. Silver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service