Skip to Content
MilliporeSigma
All Photos(4)

Documents

401668

Sigma-Aldrich

Iron(II) tetrafluoroborate hexahydrate

greener alternative

97%

Synonym(s):

Iron bis(tetrafluoroborate) hexahydrate, Iron(2+) ditetrafluoroborate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(BF4)2 · 6H2O
CAS Number:
Molecular Weight:
337.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

crystalline

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

battery manufacturing

greener alternative category

SMILES string

O.O.O.O.O.O.[Fe++].F[B-](F)(F)F.F[B-](F)(F)F

InChI

1S/2BF4.Fe.6H2O/c2*2-1(3,4)5;;;;;;;/h;;;6*1H2/q2*-1;+2;;;;;;

InChI key

NKQGOEGCKHXNEP-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for greener reduction of aldehydes to alcohols under transfer hydrogenation conditions.

Fast and selective iron-catalyzed transfer hydrogenations of aldehydes

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service