About This Item
Assay
99.99% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: cadmium
mp
232 °C (lit.)
density
4.79 g/mL at 25 °C (lit.)
SMILES string
O[Cd]O
InChI
1S/Cd.2H2O/h;2*1H2/q+2;;/p-2
InChI key
PLLZRTNVEXYBNA-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Inhalation
Target Organs
Kidney,Lungs,Bone
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service