Skip to Content
MilliporeSigma
All Photos(2)

Documents

401471

Sigma-Aldrich

Palladium(II) hexafluoroacetylacetonate

Synonym(s):

Pd(II) hexafluoroacetylacetonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(C5HF6O2)2
CAS Number:
Molecular Weight:
520.52
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder or crystals

reaction suitability

core: palladium
reagent type: catalyst

SMILES string

FC(F)(F)C(=O)\C=C(/O[Pd]O\C(=C/C(=O)C(F)(F)F)C(F)(F)F)C(F)(F)F

InChI

1S/2C5H2F6O2.Pd/c2*6-4(7,8)2(12)1-3(13)5(9,10)11;/h2*1,12H;/q;;+2/p-2/b2*2-1-;

InChI key

OVIDFVVPPRGPGR-PAMPIZDHSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used in a redox transmetalation reaction to enhance CVD alloying technology for copper. Also used in the simultaneous etching of copper and deposition of palladium for applications in ultra large-scale integrated devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin. W. et al.
Journal of the American Chemical Society, 118, 5977-5977 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service