Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

401048

Sigma-Aldrich

Cobalt(II) selenide

pieces, 6 mm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CoSe
CAS Number:
Molecular Weight:
137.89
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

pieces

particle size

6 mm

density

7.65 g/mL at 25 °C (lit.)

SMILES string

[Co]=[Se]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service