Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

400912

Sigma-Aldrich

Lithium tungstate

98%

Synonym(s):

Dilithium tetraoxotungstate, Lithium tungsten oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2WO4
CAS Number:
Molecular Weight:
261.72
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

core: tungsten
reagent type: catalyst

density

3.71 g/mL at 25 °C (lit.)

SMILES string

[Li+].[Li+].[O-][W]([O-])(=O)=O

InChI

1S/2Li.4O.W/q2*+1;;;2*-1;

InChI key

SCKKBXOJPMZVPF-UHFFFAOYSA-N

Application

  • Used for preparation of ceramics with ultra-low sintering temperatures
  • Catalyst for oxidative coupling reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service