Skip to Content
MilliporeSigma
All Photos(3)

Documents

400637

Sigma-Aldrich

Silver(I) selenide

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2Se
CAS Number:
Molecular Weight:
294.70
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

8.22 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Ag][Se][Ag]

InChI

1S/2Ag.Se

InChI key

GNWCVDGUVZRYLC-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrical properties of silver selenide thin films prepared by reactive evaporation
Kumar MCS and Pradeep B
Bulletin of Materials Science, 25(5), 407-411 null
Studies on the thermodynamic stability of silver selenide
Nasar A and Shamsuddin M
Metall. Mater. Trans. B, 28(3), 519-522 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service