Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400580

Sigma-Aldrich

Potassium tellurate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$136.00

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$136.00

About This Item

Linear Formula:
K2TeO4·xH2O
CAS Number:
Molecular Weight:
269.79 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder, crystals or chunks

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

form

powder, crystals or chunks

concentration

≤48% Te (after reduction with sulphite, gravimetric)

SMILES string

O.[K+].[K+].[O-][Te]([O-])(=O)=O

InChI

1S/2K.H2O4Te.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2

InChI key

GQDXSEHGIRKBIY-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service