Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400092

Sigma-Aldrich

(1S,2R)-(−)-2-(Dibutylamino)-1-phenyl-1-propanol

95%

Synonym(s):

(−)-DBNE, N,N-Dibutyl-D-(−)-norephedrine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2NCH(CH3)CH(C6H5)OH
CAS Number:
Molecular Weight:
263.42
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

95%

optical activity

[α]22/D −21°, c = 2 in chloroform

refractive index

n20/D 1.502 (lit.)

bp

170 °C/2 mmHg (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

CCCCN(CCCC)[C@H](C)[C@@H](O)c1ccccc1

InChI

1S/C17H29NO/c1-4-6-13-18(14-7-5-2)15(3)17(19)16-11-9-8-10-12-16/h8-12,15,17,19H,4-7,13-14H2,1-3H3/t15-,17-/m1/s1

InChI key

BRRGNOFUBFINSX-NVXWUHKLSA-N

Application

Ligand for enantioselective addition of dialkylzinc to carbonyls and imines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dae Keun Suh et al.
Cells, 11(18) (2022-09-24)
Muscle atrophy is defined as the progressive degeneration or shrinkage of myocytes and is triggered by factors such as aging, cancer, injury, inflammation, and immobilization. Considering the total amount of body iron stores and its crucial role in skeletal muscle
Soai, K.et al.
Journal of the Chemical Society. Chemical Communications, 1097-1097 (1992)
K. Soai et al.
Chemical Reviews, 92, 833-833 (1992)
Mei-Chuan Kuo et al.
Biomolecules, 11(7) (2021-08-07)
Hypoxia is regarded as one of the pathophysiologic mechanisms of kidney injury and further progression to kidney failure. Epithelial-to-mesenchymal transition (EMT) in kidney tubules is a critical process of kidney fibrosis. This study utilized transcriptome analysis to investigate hypoxia-induced EMT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service