Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3DXCFR004

Sigma-Aldrich

CarbonX CFR-PLA carbon fiber reinforced PLA 3D printing filament

black, diam. 2.85 mm, weight 750 gper each reel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.23

diam.

2.85 mm

weight

750 gper each reel

color

black

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CarbonX CFR-PLA is an improved carbon fiber reinforced 3D printing filament. This filament is ideal for anyone that desires a structrual component with high modulus, excellent surface quality, light weight, and ease of printing. Made using premium Natureworks PLA and high modulus carbon fiber.

  • 9100 Mpa Flexural Modulus (128% improvement over unfilled ABS)

Preparation Note

Recommended Print Conditions:

  • Extruder: 215 - 225 °C
  • Bed Temp: 23 - 60 °C (Heated bed NOT necessary)
  • Nozzle: minimum 0.35mm orifice recommended
  • Carbon fiber filament is more abrasive than unfilled materials; so extended use willr esult in more wear on your nozzle.

Legal Information

CarbonX is a trademark of 3DXTech

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Three-dimensional printing technology applications range from personal tools to aerospace equipment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service