Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

399841

Sigma-Aldrich

Iron(II) fluoride

98%

Synonym(s):

Ferrous fluoride, Iron difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeF2
CAS Number:
Molecular Weight:
93.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder

Assay

98%

form

powder

application(s)

battery manufacturing

SMILES string

F[Fe]F

InChI

1S/2FH.Fe/h2*1H;/q;;+2/p-2

InChI key

FZGIHSNZYGFUGM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service