Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39933

Sigma-Aldrich

(S)-4-Isopropylthiazolidine-2-thione

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NS2
CAS Number:
Molecular Weight:
161.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

optical activity

[α]20/D -37±3°, c = 1 in chloroform

optical purity

ee: ≥99:1 (LC)

mp

69-71 °C

SMILES string

CC(C)[C@H]1CSC(=S)N1

InChI

1S/C6H11NS2/c1-4(2)5-3-9-6(8)7-5/h4-5H,3H2,1-2H3,(H,7,8)/t5-/m1/s1

InChI key

CWIZUGZKLJDJLE-RXMQYKEDSA-N

Application

(S)-4-Isopropylthiazolidine-2-thione may be used as a chiral auxiliary for the stereochemical induction at the phosphorus atom during the diastereoselective synthesis of aryloxy phosphoramidate prodrugs of 3′-deoxy-2′,3′-didehydrothymidine monophosphate (d4TMP).[1]
A highly selective and efficient chiral auxiliary[2] which can be directly reduced to its corresponding aldehyde and the chiral auxiliary by reductive cleavage with diisobutylaluminum hydride.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereoselective synthesis of aryloxy phosphoramidate prodrugs of 3'-deoxy-2', 3'-didehydrothymidine monophosphate.
Roman CA, et al.
Journal of Medicinal Chemistry, 53(21), 7675-7681 (2010)
M T Crimmins et al.
Organic letters, 2(6), 775-777 (2001-02-07)
[formula: see text] Asymmetric aldol additions using chlorotitanium enolates of thiazolidinethione propionates proceed with high diastereoselectivity for the "Evans" or "non-Evans" syn product depending on the nature and amount of the base used. With (-)-sparteine as the base, selectivities of
Velazquez. F.; Olivo, H. F.
Current Organic Chemistry, 6, 303-303 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service