Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

396338

Sigma-Aldrich

Benzyl 2,2,2-trifluoroethyl sulfide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2SCH2CF3
CAS Number:
Molecular Weight:
206.23
Beilstein:
4245572
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.490 (lit.)

bp

45-46 °C/20 mmHg (lit.)

density

1.222 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)CSCc1ccccc1

InChI

1S/C9H9F3S/c10-9(11,12)7-13-6-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

ZDVBYNCJCIYCFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl 2,2,2-trifluoroethyl sulfide is a sulphur containing organic building block. Various physical properties (density and refractive index) of benzyl 2,2,2-trifluoroethyl sulfide have been reported.[1]

Application

Benzyl 2,2,2-trifluoroethyl sulfide may be used in the synthesis of 2,2,2-trifluoroethanesulfonic acid.[2]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2, 2, 2-trifluoroethanesulfonic acid.
Bunyagidj C, et al.
The Journal of Organic Chemistry, 46(16), 3335-3336 (1981)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 234-234 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service