Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

396168

Sigma-Aldrich

Strontium zirconate

powder, 10 μm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrZrO3
CAS Number:
Molecular Weight:
226.84
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

reagent type: catalyst
core: zirconium

particle size

10 μm

SMILES string

[Sr++].[O-][Zr]([O-])=O

InChI

1S/3O.Sr.Zr/q;2*-1;+2;

InChI key

FCCTVDGKMTZSPU-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service