Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

394556

Sigma-Aldrich

2,4-Dichlorophenethyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2CH2OH
CAS Number:
Molecular Weight:
191.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.566 (lit.)

bp

119 °C/0.8 mmHg (lit.)

density

1.321 g/mL at 25 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

OCCc1ccc(Cl)cc1Cl

InChI

1S/C8H8Cl2O/c9-7-2-1-6(3-4-11)8(10)5-7/h1-2,5,11H,3-4H2

InChI key

JIJGKPVJAALUQQ-UHFFFAOYSA-N

General description

2,4-Dichlorophenethyl alcohol is a phenethyl alcohol derivative. Its selective electrocatalytic dehalogenation over Pd-loaded felt cathode has been investigated.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dechlorination of chloroaromatics by electrocatalytic reduction over palladium-loaded carbon felt at room temperature.
Tsyganok AI, et al
Chemosphere, 39(11), 1819-1831 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service