Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

394327

Sigma-Aldrich

2-Chloro-4,6-dimethylaniline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(CH3)2NH2
CAS Number:
Molecular Weight:
155.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

38-40 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1cc(C)c(N)c(Cl)c1

InChI

1S/C8H10ClN/c1-5-3-6(2)8(10)7(9)4-5/h3-4H,10H2,1-2H3

InChI key

LKAQFOKMMXWRRQ-UHFFFAOYSA-N

General description

2-Chloro-4,6-dimethylaniline is an aromatic amine. It is an organic building block used in chemical synthesis. Its boiling point, freezing point, density and refractive index have been determined.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 195-195 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service