Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

394130

Sigma-Aldrich

4-Nitro-N-propylbenzamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CONHCH2CH2CH3
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

100-103 °C (dec.) (lit.)

functional group

amide
nitro

SMILES string

CCCNC(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C10H12N2O3/c1-2-7-11-10(13)8-3-5-9(6-4-8)12(14)15/h3-6H,2,7H2,1H3,(H,11,13)

InChI key

MDRVSZJDSUDYGP-UHFFFAOYSA-N

General description

4-Nitro-N-propylbenzamide ((4-nitrophenyl)-N-propylcarboxamide) is a benzamide derivative. It can be synthesized by the reaction of 4-nitrobenzoyl chloride and amine.[1]

Application

4-Nitro-N-propylbenzamide may be used in the synthesis of 4-amino-N-substituted amines by catalytic hydrogenation.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C R Clark et al.
Journal of medicinal chemistry, 27(6), 779-782 (1984-06-01)
A series of 4- aminobenzamides of some simple primary and secondary amines were prepared and evaluated for anticonvulsant effects. The compounds were tested in mice against seizures induced by electroshock and pentylenetetrazole ( metrazole ) and in the rotorod assay

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service