Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

394114

Sigma-Aldrich

2-Amino-4-phenylphenol

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(C6H5)OH
CAS Number:
Molecular Weight:
185.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

powder

mp

198-202 °C (lit.)

functional group

phenyl

SMILES string

Nc1cc(ccc1O)-c2ccccc2

InChI

1S/C12H11NO/c13-11-8-10(6-7-12(11)14)9-4-2-1-3-5-9/h1-8,14H,13H2

InChI key

IGIDZGNPFWGICD-UHFFFAOYSA-N

General description

2-Amino-4-phenylphenol is an aminoalcohol derivative. It participates in the synthesis of bisbenzoxazoles.[1]

Application

2-Amino-4-phenylphenol is suitable as an internal standard to investigate the enzymatic activity of N-hydroxy-N-2-fluorenylacetamide (N-OH-2-FAA), a mammary gland carcinogen in the female Sprague-Dawley rat, by HPLC.[2]
It may be used as a chelating agent for metal ions.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of 4, 4'-bis [5-alkyl (aryl) benzoxazol-2-yl]-2-hydroxy (alkoxy) biphenyls.
Ol'khovik VK, et al.
Russ. J. Org. Chem., 42(8), 1164-1168 (2006)
Metal ion complexing properties of carcinogen metabolites.
J H WEISBURGER et al.
Biochemical pharmacology, 12, 179-191 (1963-02-01)
C L Ritter et al.
Carcinogenesis, 17(11), 2411-2418 (1996-11-01)
N-Hydroxy-N-2-fluorenylacetamide (N-OH-2-FAA) and its benzamide analogue N-OH-2-FBA are mammary gland carcinogens in the female Sprague-Dawley rat. Ovariectomy inhibits tumorigenicity of topically applied N-OH-2-FAA suggesting modulation of carcinogen-activating enzymes in the gland. This study concerned the activation of N-OH-2-FAA and N-OH-2-FBA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service