Skip to Content
MilliporeSigma
All Photos(1)

Documents

393576

Sigma-Aldrich

3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2ClF4N
CAS Number:
Molecular Weight:
199.53
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.433 (lit.)

bp

50-55 °C/11 mmHg (lit.)

mp

16-20 °C (lit.)

density

1.524 g/mL at 25 °C (lit.)

SMILES string

Fc1ncc(cc1Cl)C(F)(F)F

InChI

1S/C6H2ClF4N/c7-4-1-3(6(9,10)11)2-12-5(4)8/h1-2H

InChI key

GDSROTVTTLUHCO-UHFFFAOYSA-N

General description

3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine, a halogenated pyridine derivative, is a fluorinated building block. It participates in the synthesis of 2,3-difluoro-5-(trifluoromethyl)pyridine.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective substitution in aromatic six-membered nitrogen heterocycles.
Comins DL and O'Connor S.
Advances in Heterocyclic Chemistry, 44, 199-267 (1988)
Catalytic fluorination of 2-chloropyridine over metal oxide catalysts in gas phase in the presence of HF.
Cochon C, et al.
Applied Catalysis A: General, 413, 149-156 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service