Skip to Content
MilliporeSigma
All Photos(1)

Documents

393223

Sigma-Aldrich

1,2-Bis(3-indenyl)ethane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18
CAS Number:
Molecular Weight:
258.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

121-125 °C (lit.)

SMILES string

C(CC1=CCc2ccccc12)C3=CCc4ccccc34

InChI

1S/C20H18/c1-3-7-19-15(5-1)9-11-17(19)13-14-18-12-10-16-6-2-4-8-20(16)18/h1-8,11-12H,9-10,13-14H2

InChI key

CQAQBIQKEFJNRZ-UHFFFAOYSA-N

General description

1,2-Bis(3-indenyl)ethane is an indene derivative.

Application

1,2-Bis(3-indenyl)ethane may be used in the synthesis of rac-(EBI)Zr(NMe2)2 (EBI=ethylene-1,2-bis(1-indenyl)) by elimination reaction with Zr(NMe2)4 and also in the preparation of dilithio-1,2-bis(indenyl)ethane. It may be used for the preparation of chiral polymerization catalysts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Synthesis of Chiral ansa-Metallocenes by Amine Elimination. Synthesis, Structure, and Reactivity of rac-(EBI)Zr(NMe2)2.
Diamond GM, et al.
Journal of the American Chemical Society, 118(34), 8024-8033 (1996)
ansa-Metallocene Derivatives: IV. Synthesis and molecular structures of chiral ansa-titanocene derivatives with bridged tetrahydroindenyl ligands.
Wild FRWP, et al.
Journal of Organometallic Chemistry, 232(3), 233-247 (1982)
Organometallics, 10, 1501-1501 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service