Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

392944

Sigma-Aldrich

Allyl chloroacetate

98%

Synonym(s):

Chloroacetic acid allyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CO2CH2CH=CH2
CAS Number:
Molecular Weight:
134.56
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

164 °C/265 mmHg (lit.)

density

1.159 g/mL at 25 °C (lit.)

SMILES string

ClCC(=O)OCC=C

InChI

1S/C5H7ClO2/c1-2-3-8-5(7)4-6/h2H,1,3-4H2

InChI key

VMBJJCDVORDOCF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service