Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

392529

Sigma-Aldrich

4-Methoxy-1-indanone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

115-120 °C/0.5 mmHg (lit.)

mp

105-107 °C (dec.) (lit.)

functional group

ketone

SMILES string

COc1cccc2C(=O)CCc12

InChI

1S/C10H10O2/c1-12-10-4-2-3-7-8(10)5-6-9(7)11/h2-4H,5-6H2,1H3

InChI key

BTYSYELHQDGJAB-UHFFFAOYSA-N

General description

4-Methoxy-1-indanone, a benzo-fused ketone[1] is a 1-indanone derivative. Its synthesis has been reported.[2]

Application

4-Methoxy-1-indanone is suitable for use in the comparative study of the effect of different substituents on various benzo-fused ketones on the reaction kinetics by studying the biocatalysed oxidation reaction.[1]
It may be used in the following studies:
  • Synthesis of isomeric mixture of oximes.[2]
  • As a starting material in the synthesis of benzo-fused indolizidine[3] and 4-methoxy-1-indanyl compound.[4]
  • Synthesis of 4-methoxy-5-nitro-1-indanone by nitration reaction.[4]
  • As one of the reactant in the synthesis of E-2-chloro-8-methyl-3-[(4′-methoxy-1′-indanoyl)-2′-methyliden]-quinoline (IQ), a quinoline derivative.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W H Pearson et al.
The Journal of organic chemistry, 65(21), 7158-7174 (2000-10-14)
The intramolecular capture of benzocyclobutyl, benzocyclopentyl, and benzocyclohexyl carbocations 7 by azides produces spirocyclic aminodiazonium ions 8, which undergo 1,2-C-to-N rearrangement with loss of dinitrogen to produce benzo-fused iminium ions resulting from either aryl (9) or alkyl (10) migration to
Juan Rodrigues et al.
Memorias do Instituto Oswaldo Cruz, 104(6), 865-870 (2009-10-31)
E-2-chloro-8-methyl-3-[(4'-methoxy-1'-indanoyl)-2'-methyliden]-quinoline (IQ) is a new quinoline derivative which has been reported as a haemoglobin degradation and ss-haematin formation inhibitor. The haemoglobin proteolysis induced by Plasmodium parasites represents a source of amino acids and haeme, leading to oxidative stress in infected
Beckmann rearrangements of 1-indanone oxime derivatives using aluminum chloride and mechanistic considerations.
Lee BS, et al.
Bull. Korean Chem. Soc., 21(9), 860-866 (2000)
Enzymatic Baeyer-Villiger Oxidation of Benzo-Fused Ketones: Formation of Regiocomplementary Lactones.
Rioz-Martinez A, et al.
European Journal of Organic Chemistry, 2009(15), 2526-2532 (2009)
Ju-Ok Lim et al.
European journal of medicinal chemistry, 44(1), 322-331 (2008-04-15)
A series of bicyclic analogues having indan and tetrahydronaphthalene templates in the A-region were designed as conformationally constrained analogues of our previously reported potent TRPV1 antagonists (1, 3). The activities for rat TRPV1 of the conformationally restricted analogues were moderately

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service