Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

390399

Sigma-Aldrich

3,5-Bis(trifluoromethyl)pyrazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2F6N2
CAS Number:
Molecular Weight:
204.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

83-85 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)c1cc([nH]n1)C(F)(F)F

InChI

1S/C5H2F6N2/c6-4(7,8)2-1-3(13-12-2)5(9,10)11/h1H,(H,12,13)

InChI key

NGDDUAYSWPUSLX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kristoffer Bernhem et al.
BMC cardiovascular disorders, 17(1), 126-126 (2017-05-19)
Blockers of angiotensin II type 1 receptor (AT To elucidate these questions, we have studied the Ca Repeated exposure to pharmacological (100 nM) concentrations of AngII caused, as expected, a down-regulation of the Ca Our findings have implications for the understanding
You-Jin Chang et al.
Experimental and therapeutic medicine, 13(2), 567-575 (2017-03-30)
In traditional Korean/Asian medicine, Salvia plebeia R.Br. (
Fumiyuki Sasaki et al.
Biochemical and biophysical research communications, 521(3), 806-813 (2019-11-12)
Osteocytes function as critical regulators of bone homeostasis by coordinating the functions of osteoblasts and osteoclasts, and are constantly exposed to mechanical force. However, the molecular mechanism underlying the mechanical signal transduction in osteocytes is not well understood. Here, we
Maren Stavermann et al.
Cell calcium, 58(2), 215-225 (2015-06-21)
Olfactory ensheathing cells (OECs) are a specialised type of glial cells, supporting axon growth and guidance during development and regeneration of the olfactory nerve and the nerve layer of the olfactory bulb. We measured calcium signalling in OECs in olfactory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service