Skip to Content
MilliporeSigma
All Photos(1)

Documents

390364

Sigma-Aldrich

[1,2-Bis(diphenylphosphino)ethane]dichloroiron(II)

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PCH2CH2P(C6H5)2]FeCl2
CAS Number:
Molecular Weight:
525.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

96%

mp

236 °C (dec.) (lit.)

SMILES string

Cl[Fe]Cl.C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service