Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

389889

Sigma-Aldrich

2,3-(Methylenedioxy)benzaldehyde

98%

Synonym(s):

1,3-Benzodioxole-4-carboxaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$71.80
5 G
$122.00

$71.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$71.80
5 G
$122.00

About This Item

Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
Beilstein:
125644
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$71.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

80 °C/0.3 mmHg (lit.)

mp

34-36 °C (lit.)

density

1.312 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1cccc2OCOc12

InChI

1S/C8H6O3/c9-4-6-2-1-3-7-8(6)11-5-10-7/h1-4H,5H2

InChI key

QZMQKPGVXNSITP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Barnali Naskar et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 20(12), 1630-1639 (2019-04-16)
Fluorescent chemosensors with aggregation induced emission enhancement (AIEE) emerge as promising tools in the field of sensing materials. Herein, we report the design, synthesis and applicability of a Schiff base chemosensor 1-(benzo[1,3]dioxol-4-ylmethylene-hydrazonomethyl)-naphthalen-2-ol (Hbdhn) of AIE characteristics that exhibits highly effective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service